missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15612780
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

for IR spectroscopy, contains ∼0.5% ethanol as stabilizer,≥99.5%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Color Colorless
Density 1.492g/mL at 25°C
Boiling Point 60°C to 62°C
Packaging Glass bottle
Refractive Index 1.446
Quantity 500 mL
UN Number UN1888
Beilstein 1731042
Vapor Density 4.1 (vs air)
Formula Weight 119.38g/mol
Vapor Pressure 160mmHg (20°C)
Percent Purity ≥99.5%
Chemical Name or Material Chloroform, Contains≈0.5% ethanol as stabilizer, for IR spectroscopy
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.