Learn More
Chloroform, 99.9%, for residue analysis, ECD tested for pesticide anal., stab. with Amylene, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -63.0°C |
Density | 1.4800g/mL |
Boiling Point | 61.0°C |
Quantity | 1 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | 99.9% |
Residue after Evaporation | 0.0005% max. |
Specific Gravity | 1.48 |
Linear Formula | CHCl3 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465291 | Certificate of Analysis | 14/08/2024 | 326670010,326670100,326670025 |
A0460224 | Certificate of Analysis | 23/02/2024 | 326670010,326670100,326670025 |
A0457077 | Certificate of Analysis | 03/11/2023 | 326670010,326670100,326670025 |
A0442998 | Certificate of Analysis | 25/08/2023 | 326670010,326670100,326670025 |
A0452675 | Certificate of Analysis | 28/04/2023 | 326670010,326670100,326670025 |
Safety and Handling


- Chloroform, stabilized with amylene
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H372-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.