Learn More
Chloroform, 99+%, Thermo Scientific Chemicals
59.50 € - 763.80 €
Chemical Identifiers
CAS | 67-66-3, 64-17-5 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformioShow More |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
IUPAC Name | chloroform |
SMILES | ClC(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071970
|
Thermo Scientific Acros
158210010 |
1 L | Glass Bottle |
59.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236110
|
Thermo Scientific Acros
158210025 |
2.5 L | Glass Bottle |
118.01 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10704661
|
Thermo Scientific Acros
158210250 |
25 L | Plastic Can |
763.80 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-66-3, 64-17-5 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
chloroform |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformioShow More | |
CHEBI:35255 | |
ClC(Cl)Cl |
Specifications
-63.0°C | |
290°C | |
61.0°C | |
1.4443 to 1.4463 | |
0.002% max. | |
CHCl3 | |
MFCD00000826 | |
0.56 mPa.s (20°C) | |
0.02% max. (K.F.) | |
15, 2142 | |
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
≥99% | |
Chloroform |
67-66-3, 64-17-5 | |
1.4800g/mL | |
Liquid | |
99+% | |
1.48 | |
CHCl3 | |
Vapor Pressure: 213mbar at 20°C | |
01, 61 | |
01,130; 08,92; 15,84 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
6212 | |
119.38 | |
Extra Pure |
Safety and Handling
GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Warning
- Chloroform, stabilized with ethanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H336-May cause drowsiness or dizziness.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Chloroform, Ethyl alcohol
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.