missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, ACS Reagent, With Stabilizer (Amylene), ≥99.8%, Honeywell Riedel-de Haën™
ACS Reagent, contains amylenes as stabilizer,≥99.8%
34.24 € - 364.82 €
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
SMILES | ClC(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15675640
|
Honeywell-Riedel-de Haen
472476-500ML |
500 mL | Glass Bottle | N/A | |||||
15655640
|
Honeywell-Riedel-de Haen
472476-1L |
1 L | Glass Bottle |
78.95 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15665640
|
Honeywell-Riedel-de Haen
472476-2.5L |
2.5 L | Glass Bottle |
173.92 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15685640
|
Honeywell-Riedel-de Haen
472476-6X1L |
6 x 1 L | Glass Bottle |
364.82 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
ClC(Cl)Cl |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 |
Specifications
67-66-3 | |
1.492g/mL at 25°C, 1.48g/mL at 25°C | |
Sweet | |
MFCD00000826 | |
1731042 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
CHEBI:35255 | |
160mmHg (20°C) | |
ACS Reagent | |
Chloroform, Containsamylenes as stabilizer |
-61.3°C | |
61°C | |
CHCl3 | |
UN1888 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
4.1 (vs air) | |
119.38g/mol | |
≥99.8% | |
Liquid |
Safety and Handling
P261-P281-P305 + P351 + P338-P311
missing translation for 'dotInformation' : 6.1/PGIII
missing translation for 'rtecsNumber' : 200-663-8
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title