missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Description
- For Histological Use
Specifications
Specifications
Quantity | 1 L |
Packaging | Aluminum Bottle |
Specific Gravity | 1L = 1.48kg |
Formula Weight | 119.38 |
Grade | Meets ACS reagent chemical requirements |
Chemical Name or Material | Chloroform |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P281-Use personal protective equipment as required.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'