missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
50 mL
100 mL
250 mL
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Specifications
Physical Form | Crystals or Crystalline Powder |
Color | White |
Density | 1.5000g/mL |
Deuterium (D) | 99.75% min. |
Packaging | Glass Bottle |
Water | 0.02% max. (Coulometric) |
Quantity | 50 mL |
Melting Point | -64.0°C |
Boiling Point | 60.0°C |
Linear Formula | CDCl3 |
Show More |
Safety and Handling
Product Identifier
- Chloroform-d
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'