missing translation for 'onlineSavingsMsg'
Learn More
Chloroform, for HPLC, Stabilised with Amylene, Fisher Chemical Product Code.: 10427060

Chloroform, for HPLC, Stabilised with Amylene, Fisher Chemical

Product Code. 10427060
2.5L, Plastic Coated Glass bottle
Click to view available options
2.5L, Plastic Coated Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10427060

Brand: Fisher Chemical C/4966/PB17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

Melting Point -63°C
Color Colorless
Boiling Point 61°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Vapor Pressure 213mbar at 20°C
Viscosity 0.56 mPaS at 20°C
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with amylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroform, for HPLC, Stabilised with Amylene, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.