missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Hydrocarbone Stabilized, BAKER ANALYZED HPLC, J.T. Baker™
Get rapid, reproducible performance and separation results. Chloroform, Hydrocarbone Stabilized, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.
104.00 € - 256.00 €
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
SMILES | ClC(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10140591
|
JT Baker
9174.1000 |
1 L |
104.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10078140
|
JT Baker
9174.2500 |
2.5 L |
256.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.
- Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
- Low backgrounds free of extraneous peaks
- Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
- Fluorescence testing for trace impurities which would cause interference
- Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
- Lot to lot consistency, with an actual Lot Analysis on the label
- Innovative packaging options to assure solvent quality to the point-of-use
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
ClC(Cl)Cl |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 |
Specifications
-63°C | |
67-66-3 | |
1.483g/mL at 20°C | |
982°C | |
Glass | |
Max. 5 ppm | |
Max. 0.0005meq/g | |
0.56 mPas at 20°C | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
119.38g/mol | |
BAKER ANALYZED™ HPLC |
Max. 0.15 at 254nm, Max. 0.05 at 280nm, Max. 0.01 at 350nm | |
Colorless | |
62°C | |
Sweetish | |
Liquid | |
CHCl3 | |
MFCD00000826 | |
Max. 0.01% | |
8g/L in water | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
≥99.8% | |
Chloroform |
Safety and Handling
missing translation for 'einecsNumber' : 200-663-8
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P281-Use personal protective equipment as required.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title