missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform Molecular Biology MP Biomedicals
Click to view available options
Quantity:
25 mL
100 mL
0.5 L
1 L
Specifications
Specifications
Density | 4.36 (air = 1) |
Quantity | 25 mL |
IUPAC Name | chloroform |
Formula Weight | 119.38 |
Grade | Molecular Biology |
Chemical Name or Material | Chloroform |
Safety and Handling
Product Identifier
- Chloroform, Molecular Biology Reagent
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : chloroform
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'