missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform Molecular Biology MP Biomedicals
Click to view available options
Quantity:
25 mL
100 mL
0.5 L
1 L
Specifications
Specifications
Density | 4.36 (air = 1) |
Quantity | 100 mL |
IUPAC Name | chloroform |
Formula Weight | 119.38 |
Grade | Molecular Biology |
Chemical Name or Material | Chloroform |
Safety and Handling
Product Identifier
- Chloroform, Molecular Biology Reagent
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction