missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Reagent ACS, 99.8+%, Thermo Scientific Chemicals

Product Code. 10311551
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3, 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.0°C
Decomposition Information 290°C
Density 1.4800g/mL
Boiling Point 61.0°C
Quantity 25 mL
Infrared Spectrum Conforms to Ref Report
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4443 to 1.4463
Assay Percent Range 99.8+%
Residue after Evaporation ≤0.001%
Specific Gravity 1.48
Linear Formula CHCl3
Vapor Pressure Vapor Pressure: 213mbar at 20°C
Viscosity 0.56 mPa.s (20°C)
Beilstein 01, 61
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
IUPAC Name chloroform
Formula Weight 119.38
Percent Purity ≥99.8%
Grade ACS Reagent
Chemical Name or Material Chloroform, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with ethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H336-May cause drowsiness or dizziness.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Chloroform, Ethyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.