missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals

Product Code. 10289473
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chloroform, >-99%, CHCl3, CAS Number-67-66-3, 64-17-5, trichloromethane, trichloroform, trichlormethan, methyl trichloride, cloroformio, methenyl trichloride, formyl trichloride, methane, trichloro, chloroforme, methane trichloride, 2.5L

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3, 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.0°C
Absorbance 0.009 max. at 300nm, 0.046 max. at 265nm, 0.301 max. at 250nm, 0.699 max. at 245nm, 1 cm cell vs water, 1 cm cell vs water at 0nm
Color White
Density 1.4800g/mL
Boiling Point 61.0°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Crystals
Refractive Index 1.4443 to 1.4463
Assay Percent Range 99+%
Residue after Evaporation 0.0002 % max.
Specific Gravity 1.48
Linear Formula CHCl3
Viscosity 0.56 mPa.s (20°C)
Water 0.01% Max. (K.F.)
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
IUPAC Name trichloromethane
Formula Weight 119.38
Percent Purity ≥99%
Grade HPLC
Chemical Name or Material Chloroform, Stabilized with ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'