missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Technical, amylene stabilized, ≥99% (GC), Honeywell

Product Code. 15612600
Click to view available options
Quantity:
250 kg
Packaging:
Metal Drum
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Technical, amylene stabilized,≥99% (GC)

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-66-3
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Melting Point -63°C (literature)
Density 1.492 g/mL at 25 °C(lit.)
Boiling Point 61°C
Packaging Metal Drum
Molecular Formula CHCl3
Linear Formula C2H5COCH3
MDL Number MFCD00000826
Quantity 250 kg
UN Number UN1888
Beilstein 1731042
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name chloroform
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38
Percent Purity >99%
Grade Technical
Chemical Name or Material Chloroform, Technical, amylene stabilized, ≥99% (GC)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.