missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl chloroformate, 97%, Thermo Scientific Chemicals

64.62 € - 231.15 €

Chemical Identifiers

CAS 22128-62-7
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00077688
InChI Key JYWJULGYGOLCGW-UHFFFAOYSA-N
Synonym chloromethyl chloroformate, chloromethylchloroformate, chloroformic acid chloromethyl ester, carbonochloridic acid, chloromethyl ester, palite, chlormethylchlorformiat, clco2ch2cl, chlormethylchlorocarbonat, chloromethyl choroformate, choromethyl chloroformate
PubChem CID 62754
IUPAC Name chloromethyl carbonochloridate
SMILES C(OC(=O)Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317476
View Documents
Thermo Scientific Alfa Aesar
L09527.09
10 g
64.62 €
10g
Please to purchase this item. Need a web account? Register with us today!
11327476
View Documents
Thermo Scientific Alfa Aesar
L09527.18
50 g
231.15 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents and water.
Specifications

Chemical Identifiers

22128-62-7
128.936
JYWJULGYGOLCGW-UHFFFAOYSA-N
62754
C(OC(=O)Cl)Cl
C2H2Cl2O2
MFCD00077688
chloromethyl chloroformate, chloromethylchloroformate, chloroformic acid chloromethyl ester, carbonochloridic acid, chloromethyl ester, palite, chlormethylchlorformiat, clco2ch2cl, chlormethylchlorocarbonat, chloromethyl choroformate, choromethyl chloroformate
chloromethyl carbonochloridate

Specifications

22128-62-7
1.465
95°C (203°F)
C2H2Cl2O2
1.4295
UN2745
Moisture sensitive
Slightly miscible with water.
C(OC(=O)Cl)Cl
128.936
128.94
Chloromethyl chloroformate
<-20°C
107°C to 108°C
Acrid
ClCO2CH2Cl
MFCD00077688
506426
chloromethyl chloroformate, chloromethylchloroformate, chloroformic acid chloromethyl ester, carbonochloridic acid, chloromethyl ester, palite, chlormethylchlorformiat, clco2ch2cl, chlormethylchlorocarbonat, chloromethyl choroformate, choromethyl chloroformate
JYWJULGYGOLCGW-UHFFFAOYSA-N
chloromethyl carbonochloridate
62754
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H330-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROMETHYL CHLOROFORMATE

missing translation for 'einecsNumber' : 244-793-3

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.