missing translation for 'onlineSavingsMsg'
Learn More

Chloromethylphosphonic acid, 98%, Thermo Scientific Chemicals

Product Code. 15254874
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2565-58-4
Molecular Formula CH4ClO3P
Molecular Weight (g/mol) 130.46
MDL Number MFCD00013955
InChI Key MOFCYHDQWIZKMY-UHFFFAOYSA-N
Synonym chloromethyl phosphonic acid, phosphonic acid, chloromethyl, chloromethanephosphonic acid, phosphonic acid, p-chloromethyl, monochloromethyl phosphonic acid, chloromethylphosphite, phosphonomethyl chloride, chloromethyl-phosphonicaci, phosphonic acid,p-chloromethyl, chloromethyl phosphonic acid, 98 %
PubChem CID 75723
IUPAC Name chloromethylphosphonic acid
SMILES OP(O)(=O)CCl

Specifications

Melting Point 88.0°C to 92.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 130.47
Percent Purity 98%
Chemical Name or Material Chloromethylphosphonic Acid
compliance-icons
Product Identifier
  • Chloromethylphosphonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'