missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloropentacarbonylrhenium(I), 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
Pentacarbonylchlororhenium(I) was used to study π-electron deficient complexes between tricarbonylhalorhenium and diimine ligands a well as to prepare [3]-ferrocenophane complexes. It has also been employed in preparation of a series of mixed-valence compounds of rhenium and iron. These compounds find application in the study of charge-transfer processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 14099-01-5 |
Color | Brown to White |
Molecular Formula | C5ClO5Re |
Linear Formula | Re(CO)5Cl |
MDL Number | MFCD00013296 |
Quantity | 1 g |
UN Number | UN3466 |
Sensitivity | Air and moisture sensitive |
Synonym | pentacarbonylchlororhenium, carbon monoxide; chlororhenium, pentakis carbon monoxide ; chlororhenium, 5co.clre, rhenium i pentacarbonyl chloride, pentacarbonylchlororhenium i |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'