missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriethylsilane, 99%

Product Code. 10032870 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10032870

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 994-30-9 | C6H15ClSi | 150.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-30-9
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00000507
InChI Key DCFKHNIGBAHNSS-UHFFFAOYSA-N
Synonym triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
PubChem CID 13819
IUPAC Name chloro(triethyl)silane
SMILES CC[Si](Cl)(CC)CC

Specifications

Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 142.0°C to 144.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)3SiCl
Refractive Index 1.4300 to 1.4320
Quantity 100 mL
Beilstein 04,627
Fieser 13,165
Merck Index 15,2179
Specific Gravity 0.89
Solubility Information Solubility in water: decomposes
Formula Weight 150.72
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Chlorotriethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorotriethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.