missing translation for 'onlineSavingsMsg'
Learn More

Chloroxynil, 97+%, Thermo Scientific Chemicals

Product Code. 11385459
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chloroxynil is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroxynil is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1891-95-8
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002177
InChI Key YRSSHOVRSMQULE-UHFFFAOYSA-N
Synonym chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile
PubChem CID 74685
IUPAC Name 3,5-dichloro-4-hydroxybenzonitrile
SMILES OC1=C(Cl)C=C(C=C1Cl)C#N

Specifications

Melting Point 143°C to 144°C
Quantity 5 g
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 188.01
Percent Purity ≥97%
Chemical Name or Material Chloroxynil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'