missing translation for 'onlineSavingsMsg'
Learn More

Chloroxynil, 97+%, Thermo Scientific Chemicals

151.00 € - 648.00 €

Chemical Identifiers

CAS 1891-95-8
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002177
InChI Key YRSSHOVRSMQULE-UHFFFAOYSA-N
Synonym chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile
PubChem CID 74685
IUPAC Name 3,5-dichloro-4-hydroxybenzonitrile
SMILES OC1=C(Cl)C=C(C=C1Cl)C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11375459
View Documents
Thermo Scientific Alfa Aesar
H50432.03
1 g
151.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11385459
View Documents
Thermo Scientific Alfa Aesar
H50432.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Chloroxynil is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroxynil is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications

Chemical Identifiers

1891-95-8
188.01
YRSSHOVRSMQULE-UHFFFAOYSA-N
74685
OC1=C(Cl)C=C(C=C1Cl)C#N
C7H3Cl2NO
MFCD00002177
chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile
3,5-dichloro-4-hydroxybenzonitrile

Specifications

1891-95-8
C7H3Cl2NO
UN3439
Slightly soluble in water.
OC1=C(Cl)C=C(C=C1Cl)C#N
188.01
188.01
Chloroxynil
143°C to 144°C
MFCD00002177
chloroxynil, benzonitrile, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzonitrile, chloroxynil iso, caswell no. 309a, unii-gi6x21wsvn, gi6x21wsvn, epa pesticide chemical code 309500, 4-cyano-2,6-dichlorophenol, 3,5-dichloro-4-hydroxy benzonitrile
YRSSHOVRSMQULE-UHFFFAOYSA-N
3,5-dichloro-4-hydroxybenzonitrile
74685
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 217-572-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only