missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl Chloroformate 97%, Thermo Scientific Chemicals

Product Code. 10429020
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7144-08-3
Molecular Formula C28H45ClO2
Molecular Weight (g/mol) 449.12
MDL Number MFCD00003633
InChI Key QNEPTKZEXBPDLF-JDTILAPWSA-N
Synonym cholesteryl chloroformate, cholesterol chloroformate, cholesterol, chloroformate, cholesteryloxycarbonyl chloride, cholest-5-en-3-ol 3.beta.-, carbonochloridate, 1r,3as,3bs,7s,9ar,9bs,11ar-9a,11a-dimethyl-1-2r-6-methylheptan-2-yl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl carbonochloridate, cholesterol chlorocarbonate, chlolesterol chloroformate, cholesterol, chloroformate 8ci
PubChem CID 111262
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] carbonochloridate
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)Cl)C)C

Specifications

Melting Point 116°C to 118°C
Color White
Loss on Drying 0.2% max. (60°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Fieser 01,141
Specific Rotation -26° to -30° (27°C, 589nm) (c=2, CHCl3)
Formula Weight 449.1
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Cholesteryl chloroformate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Cholesteryl chloroformate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.