missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CHPG Sodium salt
Selective mGlu5 agonist. Sodium salt of CHPG (Cat. No. 1049)
193.00 € - 758.00 €
Specifications
Purity | >99% |
---|---|
Product Type | CHPG Sodium salt |
Molecular Weight (g/mol) | 223.59 |
Inhibitors | CHPG Sodium salt |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18005272
|
Tocris Bioscience™
3695/10 |
10 mg |
193.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18004302
|
Tocris Bioscience™
3695/50 |
50 mg |
758.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium salt of the selective mGlu5 agonist CHPG (Cat. No. 1049).Specifications
>99% | |
CHPG Sodium salt | |
223.59 | |
CHPG Sodium salt |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title