Learn More
Christeyns™ Chriox Spray
Chriox spray is an ecological, ready-to-use disinfectant based on peracetic acid.
Brand:  Christeyns™ 391172
 View more versions of this product
Product Code. 17890245
  • 45.00 € / 4.50 L
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Eco-detergent certified by ECOCERT Greenlife according to the ECOCERT standard
  • Suitable for cleaning all washable surfaces (including in the food industry)
  • Spray the pure product on the surface to be treated or preferably on a clean, nonwoven cloth, and leave on for 5 to 15 min.
Specifications
Specifications
Surface Disinfectant
Clear Liquid
Unscented
6 Bottles
No
3 to 3.4
Non-sterile
750 mL