missing translation for 'onlineSavingsMsg'
Learn More

CHRNA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55291-1-AP-150UL

 View more versions of this product

Product Code. 16826785

  • 410.00 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody is specific to CHRNA10.

Ionotropic receptor with a probable role in the modulation of auditory stimuli. Agonist binding may induce an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane. The channel is permeable to a range of divalent cations including calcium, the influx of which may activate a potassium current which hyperpolarizes the cell membrane. In the ear, this may lead to a reduction in basilar membrane motion, altering the activity of auditory nerve fibers and reducing the range of dynamic hearing. This may protect against acoustic trauma.
Specifications

Specifications

CHRNA10
Polyclonal
Unconjugated
CHRNA10
CHRNA10, NACHR alpha 10, NACHRA10
Rabbit
Antigen Affinity Chromatography
RUO
504186, 57053, 64574
-20°C
Liquid
Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9GZZ6, Q9JLB5
Chrna10
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers