missing translation for 'onlineSavingsMsg'
Learn More

Chroma Gesellschaft™ 13mm Plier Style Decapper

Brand:  JG Finneran Associates 9310-13

Product Code. 10701822

  • 258.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Specifications

Specifications

11 mm
Plier-style Decapper
11 mm Caps
Crimp Top Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chroma Gesellschaft™ 13mm Plier Style Decapper >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.