missing translation for 'onlineSavingsMsg'
Learn More
Ansell™ Microflex™ 93-260 Chemical-Resistant Disposable Glove Product Code.: 15623074
Estimated Shipment: 19-05-2025
to see stock.
Add to basket

Ansell™ Microflex™ 93-260 Chemical-Resistant Disposable Glove

Product Code. 15623074
X-Small
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy

Product Code. 15623074

Brand: Ansell™ 93260060

Estimated Shipment: 19-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Provides thin yet tough chemical protection with unparalleled comfort. Microflex™ 93-260 Disposable Gloves have a solvent resistant exterior, an acid and base resistant middle layer and an easy donning interior layer.

  • Three layer design for superior protection against harsh chemicals including acids, solvents and bases
  • Thin mil construction provides enhanced tactility and dexterity
  • Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
  • Lower acceptable pinhole rate (0.65 AQL) and extended cuff for reliable protection against hazardous substances
  • Silicone free formulation and processing ensure better product protection

SIZE CHART

  • XS – 5.5 to 6
  • S – 6.5 to 7
  • M – 7.5 to 8
  • L – 6.5 to 9
  • XL – 9.5 to 10
  • 2XL – 10.5 to 11
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D3577, TYPE II, PPE 89/686, EN 420, 388 & 374 CATEGORY III
Color Green, Blue
Disposable Single-use
Hand Ambidextrous
Material Nitrile, Neoprene
Powdered Powder Free
Product Type Disposable Gloves
Size X-Small
Thickness (Metric) 0.198 mm
Finger Style Textured
Glove Cuff Style Extended, Beaded
Model 93-260
Packaging Type Dispenser box
Puncture Rating 0.65 AQL
Quantity 50Pack
Resistant To Harsh chemicals including acids, solvents and bases
Tensile Strength 22 MPa
Length (Metric) 300 mm
Width (Metric) 78 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ Microflex™ 93-260 Chemical-Resistant Disposable Glove >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.