Learn More
Minigrip™ Resealable Write-on Bags
Label bag contents with ease. Minigrip™ Resealable Write-on Bags provide three white bars specifically designed for writing on with inkpen or marker.
Brand: Minigrip™ BAJ-340-091N
Description
- Polyethylene
- Resealable
- Autoclavable
- Three write-on bars for labelling
Specifications
Resealable Bag | |
Ideal for storing stationery and sending samples | |
100 Pack | |
190 mm |
Clear | |
Polyethylene | |
190 mm |