missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ C18 Polypropylene SPE Column

Brand:  Macherey-Nagel™ 730400

 View more versions of this product

Product Code. 10489361

  • 295.00 € / Pack of 20
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Silica
45 mL
5 g
45 μm
Solid Phase Extraction Column
60 μm
14%
SPE
Solid Phase Extraction Non-Polar Compounds
Chromabond C18
No
500 m2/g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers