Learn More
Macherey-Nagel™ CHROMABOND™ Flash RS SiOH Cartridges
Ideal for Flash separations from 10mg up to 300g. Macherey-Nagel™ CHROMABOND™ Flash RS SiOH Cartridges are pre-packed columns for direct use in Teledyne Isco™ systems. Available in a range of sizes with 25–300g of SiOH adsorbent.
193.00 € - 461.00 €
Specifications
Material Polypropylene
Style Female Luer lock inlet, male Luer outlet
Type Flash RS SiOH Column
Column Type Flash Column
For Use With (Application) Flash Chromatography
View More Specs
Products 4
Product Code Brand Includes Length (Metric) Diameter (Metric) Outer Max. Pressure Price Quantity & Availability  
Product Code Brand Includes Length (Metric) Diameter (Metric) Outer Max. Pressure Price Quantity & Availability  
11653109
View Documents
Macherey-Nagel™
732802
25 g SiOH 165 mm 21.2 mm ID 21 bar
193.00 €
Pack of 15
In Stock
to see stock.
Add to basket
12768322
View Documents
Macherey-Nagel™
732806
200 g SiOH 200 mm 60.0 mm ID 12 bar
461.00 €
Pack of 6
In Stock
to see stock.
Add to basket
12853961
View Documents
Macherey-Nagel™
732805
120 g SiOH 255 mm 36.0 mm ID 15 bar
319.00 €
Pack of 10
In Stock
to see stock.
Add to basket
12758322
View Documents
Macherey-Nagel™
732804
80 g SiOH 240 mm 30.8 mm ID 15 bar
320.00 €
Pack of 12
In Stock
to see stock.
Add to basket
Description
Description
For convenient operation and reliable upscaling, these ready-to-use Flash cartridges can be used with the Isco Companion™ and other Teledyne Isco CombiFlash™ systems, or as a stand-alone version for all pump/detector combinations (e.g., from Biotage™, Büchi™)
  • Economical, ready to use cartridges increase flexibility and save time and money
  • Increases analytical safety—high pressure stability, excellent separation efficiency, good reproducibility
  • Packed with 25g to 200g SiOH adsorbent
  • Corresponding TLC plates: silica layers
  • Macherey-Nagel™ is one of the leading companies in silica and TLC
Specifications
Specifications
Polypropylene
Flash RS SiOH Column
Flash Chromatography
Female Luer lock inlet, male Luer outlet
Flash Column
Directly Used in The Teledyne Isco Companion, Rf
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ CHROMABOND™ Flash RS SiOH Cartridges