Learn More
Macherey-Nagel™ Chromabond™ HR-XAW Polypropylene SPE Column
79.05 € - 264.00 €
Specifications
Base Material Spherical PS/DVB Copolymer
Column Type SPE
Particle Size 85 μm
Sorbent Chromabond HR-XAW
Type Solid Phase Extraction Column
View More Specs
Products 7
Product Code Brand Column Capacity (Metric) Pore Size Description Price Quantity & Availability  
Product Code Brand Column Capacity (Metric) Pore Size Description Price Quantity & Availability  
12757720
View Documents
Macherey-Nagel™
730729
1 mL 55 to 65 μm Adsorbent Weight: 100mg
104.00 €
Pack of 30
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
12798192
View Documents
Macherey-Nagel™
730728
1 mL 55 to 65 Å Adsorbent Weight: 30mg
79.05 €
Pack of 30
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
12788212
View Documents
Macherey-Nagel™
730748
3 mL 55 to 65 Å Adsorbent Weight: 200mg
128.00 €
Pack of 30
In Stock
to see stock.
Add to basket
12748212
View Documents
Macherey-Nagel™
730744
3 mL 55 to 65 μm Adsorbent Weight: 500mg
186.00 €
Pack of 30
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
12768212
View Documents
Macherey-Nagel™
730747
3 mL 55 to 65 μm Adsorbent Weight: 60mg
114.00 €
Pack of 30
In Stock
to see stock.
Add to basket
12777730
View Documents
Macherey-Nagel™
730749
6 mL 55 to 65 μm Adsorbent Weight: 150mg
233.00 €
Pack of 30
In Stock
to see stock.
Add to basket
12758212
View Documents
Macherey-Nagel™
730745
6 mL 55 to 65 μm Adsorbent Weight: 500mg
264.00 €
Pack of 30
In Stock
to see stock.
Add to basket
Specifications
Spherical PS/DVB Copolymer
85 μm
Solid Phase Extraction Column
SPE
Chromabond HR-XAW
850 m2/g