missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChromaCare™ LC-MS Biologics Flush Solution, Thermo Scientific™
Flush solution for cleaning blood, plasma, and urine samples from the chromatograph.
197.00 €
Specifications
CAS | 67-64-1, 75-05-8 |
---|---|
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
Percent Purity | Pass Test (By GC-FID) |
Grade | LC-MS |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15793467
|
Thermo Scientific
MB124-212 |
2.5 L | Amber Glass Bottle | N/A | |||||
Description
- Suitable for flushing difficult to clean samples such as blood, plasma, and urine after vitamin D, hormone or drugs of abuse testing
- Save time by using off-the-shelf blend — premade for convenience
- Helps to reduce sample carryover
- Packaged in 2.5L amber glass bottle
Specifications
67-64-1, 75-05-8 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
Pass Test (By GC-FID) | |
LC-MS |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 45% Isopropyl Alcohol + 45% Acetonitrile + 10% Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile, Isopropyl alcohol, Acetone
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title