missing translation for 'onlineSavingsMsg'
Learn More

ChromaCare™ LC-MS Instrument Flush Solution

Product Code. p-8481023 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Borosilicate glass bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 15893187

Brand: Thermo Scientific T111101000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ChromaCare™ LC-MS Instrument Flush Solution is designed to prepare LC/MS instruments for start-up. By reducing background noise, this solution facilitates instrument installation and preventative maintenance routines. ∣ CAS: 75-05-8; 67-56-1; 67-63-0; 7732-18-5

  • Designed by Thermo Scientific to maximize cleaning while preserving instrument integrity in Vanquish™ and UltiMate™ UHPLC systems with QQQ MS
  • Comprised of a unique blend of polar protic and aprotic solvents for removing a wide variety of contaminants
  • Particularly suitable for new LC/MS and UHPLC/MS installations to rapidly stabilize baseline
  • A blend of 25% acetonitrile, 25% methanol, 25%, water, and 25% 2-propanol (IPA)
  • Offered in 1L borosilicate bottles and 2.5L amber glass bottles
TRUSTED_SUSTAINABILITY

Specifications

Description LC-MS Instrument Flush Solution
Packaging Borosilicate glass bottle
CAS 75-05-8; 67-56-1; 67-63-0; 7732-18-5
Physical Form Liquid
Color Colorless
Quantity 1 L
Percent Purity 25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA)
Grade LC-MS
compliance-icons compliance-icons compliance-icons
Product Identifier
  • ChromaCare LC/MS Instrument Flush Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Acetonitrile, Methanol, isopropanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.