missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chromacol Economy Septa for GC Instruments, 9mm economy GC septa

Thermo Scientific™ Chromacol Economy Septa withstand temperatures up to 250°C.

Brand:  Thermo Scientific™ ECO-9.0

Product Code. 11543670

  • 40.64 € / Pack of 25
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Chromacol Economy Septa withstand temperatures up to 250°C.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

9mm Economy GC Septa
Chromacol
Septa
9 mm
Certificate of Analysis (COA)
GC Instrument, 9 mm Economy GC Septa
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers