Learn More
Macherey-Nagel™ Chromafix™ C18 Cartridge
Brand:  Macherey-Nagel™ 731802
 View more versions of this product
Product Code. 10628621
  • 202.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Silica
480 mg
45 μm
Chromafix C18 Cartridge
50 Pack
14%
Non-Polar Compounds, Pesticides
C18
60 Å
500 m2/g