Learn More
Macherey-Nagel™ Chromafix™ C4 Cartridge
Brand:  Macherey-Nagel™ 731741
Product Code. 10092821
  • 202.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Silica
Compounds, Which Are Too Strongly Retained on C18 or C8
Chromafix C4 Cartridge
60 Å
500 m2/g
7%
45 μm
No
50 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Chromafix™ C4 Cartridge > Adsorbent Weight: 440mg