Learn More
Macherey-Nagel™ Chromafix™ HR-XA Cartridge
Brand:  Macherey-Nagel™ 731769
 View more versions of this product
Product Code. 12804441
  • 259.00 € / Pack of 50
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Spherical PS/DVB Copolymer
45 or 85 μm
Chromafix HR-XA Cartridge
50 Pack
240 mg
Strong quat. ammonium modified polymeric anion exchanger
55 to 65 Å
850 m2/g