Learn More
Macherey-Nagel™ Chromafix™ HR-XAW Cartridge
Brand:  Macherey-Nagel™ 731772
 View more versions of this product
Product Code. 12778252
  • 259.00 € / Pack of 50
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Spherical PS/DVB Copolymer
45 or 85 μm
Chromafix HR-XAW Cartridge
50 Pack
240 mg
Weakly basic secondary and tertiary ammonium polymeric anion exchanger
55 to 65 Å
850 m2/g