Learn More
Macherey-Nagel™ Chromafix™ HR-XC Cartridge
Brand:  Macherey-Nagel™ 731755
 View more versions of this product
Product Code. 12830961
  • 233.00 € / Pack of 50
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Spherical PS/DVB Copolymer
Basic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Fungicides From Food
Benzenesulfonic acid modified polymeric cation exchanger
65 to 75 Å
800 m2/g
155 mg
45 or 85 μm
Chromafix HR-XC Cartridge
50 Pack