Learn More
Macherey-Nagel™ Chromafix™ HR-XCW Cartridge
Brand:  Macherey-Nagel™ 731774
 View more versions of this product
Product Code. 12798252
  • 233.00 € / Pack of 50
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Spherical PS/DVB Copolymer
45 or 85 μm
Chromafix HR-XCW Cartridge
50 Pack
155 mg
Weak carboxylic acid modified polymeric cation exchanger
50 to 60 Å
850 m2/g