Learn More
Macherey-Nagel™ Chromafix™ PS-Ba2+ cartridges
Brand:  Macherey-Nagel™ 731871
 View more versions of this product
Product Code. 10308601
  • 233.00 € / Pack of 50
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Strong PS/DVB Cation Exchanger, Ba2+ form
Removal of Sulfate Ions From Water
Chromafix PS-Ba2+ Cartridge
50 Pack
280 mg
100 μm
100 Å