Learn More
Macherey-Nagel™ Chromafix™ PS-H+ SPE Cartridge
For removal or concentration of cations from water, decreasing the pH value of basic samples for standard protocol. Macherey-Nagel™ Chromafix™ PS-H+ SPE Cartridge is a polymer-based SPE phase for RP and ion chromatography.
Brand:  Macherey-Nagel™ 731863
 View more versions of this product
Product Code. 12471718
  • 332.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
These polymer SPE phases are used for the removal of interfering compounds and enrichment of the analytes in reverse phase and ion chromatography.
  • Strong PS/DVB cation exchanger, with H+ form capacity of 2.9 meq/g
  • Very low degree of swelling, thus very well suited for chromatography
  • Reliable function over the whole pH range from 0–14
  • Improves chromatographic separation if the interfering components overlap with the analyte
  • Improves lifetime of the column, since interfering components can block the packing
Specifications
Specifications
Strong PS/DVB Cation Exchanger, H+ form
Removal or Concentration of Cations From Water Decreasing The pH Value of Basic Samples
Chromafix PS-H+ Cartridge
50 Pack
900 g
100 μm
100 Å
L