Learn More
Macherey-Nagel™ Chromafix™ PS-RP cartridges
Brand:  Macherey-Nagel™ 731875
Product Code. 10105141
  • 251.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Hydrophobic PS/DVB Copolymer
100 μm
100 Å
Removal of Interfering Organic Components From Water
Chromafix PS-RP Cartridge
50 Pack