Learn More
Macherey-Nagel™ Chromafix™ SA Cartridge
Brand:  Macherey-Nagel™ 731832
 View more versions of this product
Product Code. 10135091
  • 210.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Silica
Amino Acids, Amines, Chlorophyll, PCBs
SA
60 Å
500 m2/g
Adsorbent Weight: 450 mg
45 μm
Chromafix SA Cartridge
50 Pack