Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and NS Socket
Brand: Lenz Laborglasinstrumente™ 05450704
This item is not returnable.
View return policy
Specifications
Chromatography Column | |
20 mm | |
125 mL | |
Duran Glass | |
0 to 2.5mm Bore PTFE-Hahn Stopcock |
Chromatographic applications | |
Chromatography Column | |
400 mm | |
20 mm |