Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and NS Socket
Brand: Lenz Laborglasinstrumente™ 05450706
This item is not returnable.
View return policy
Specifications
Chromatography Column | |
30 mm | |
430 mL | |
Duran Glass | |
0 to 2.5mm Bore PTFE-Hahn Stopcock |
Chromatographic applications | |
Chromatography Column | |
600 mm | |
30 mm |