Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and NS Socket
Brand:  Lenz Laborglasinstrumente™ 05450703
 View more versions of this product
Product Code. 10529581
  • 77.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Chromatography Column
15 mm
35 mL
Duran Glass
0 to 2.5mm Bore PTFE-Hahn Stopcock
Chromatographic applications
Chromatography Column
200 mm
15 mm