Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Indentations and Beaded Rim
Brand:  Lenz Laborglasinstrumente™ 05435704
 View more versions of this product
Product Code. 10707901
  • 83.00 € / Each
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Chromatography Column
20 mm
125 mL
Duran Glass
PTFE Stopcock and Retaining Device
Chromatographic applications
Chromatography Column
400 mm
20 mm