missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Chromatography Papers, Sheets, Grade FN 30
Highly pure paper for blotting, chromatography, and a wide range of life science and diagnostics absorption applications.
Brand: Sartorius Stedim Biotech™ FT-2-526-254305N
This item is not returnable.
View return policy
Description
- Highly pure paper grade.
- Ideal for blotting and chromatography.
- Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Non-sterile, bulk packed.
Specifications
100/Pk, 320 g/m2 | |
0.9 mm | |
FN 30 | |
305 mm |
254 mm × 305 mm | |
Sheet | |
Cotton Linters (> 98 % alpha-cellulose; < 0.04 % ash) | |
254 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction