Learn More
Macherey-Nagel™ Chrombond™ FM 15/2 SiOH Polypropylene Flash Column
For automated systems or vacuum manifolds
Brand: Macherey-Nagel™ 730881
This item is not returnable.
View return policy
Description
- Automated systems or CHROMABOND™ vacuum manifolds
- Polypropylene column, polyethylene filter element
- High loading capacity and separation efficiency
- Constant back pressure, airtight sealing
- Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
90 mm | |
Flash Column | |
Biotage FlashMaster Systems |
Polypropylene | |
Flash Column |