missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Nichrome Inoculation Loops
Click to view available options
Diameter (Metric) Inside:
1.59 mm
2.26 mm
2.52 mm
2.91 mm
3.57 mm
5.05 mm
Gauge:
Heavy
Standard
Unit Size:
Pack of 25
Descripción
An inoculation loop is a simple tool used to gather and transfer microscopic organisms from a culture or broth plate. The small loop removes a consistent amount of inoculum by using surface tension. The inoculation loop is flame-sterilized before and after each use to eliminate cross-contamination between cultures. (The loop must be cooled after sterilization to avoid destroying the organisms.)
- Constructed of economical nickel-chrome in standard or heavy gauge wire
- Available in a range of 6 sizes, 1μL / 1.59mmD to 10μL / 5.05mmD
- Packaged in sets of 25 loops
- Holders with insulated handles sold separately
Especificaciones
Especificaciones
| Volume | 3.3 μL |
| Diameter (Metric) Inside | 2.91 mm |
| Material | Chrome-Nickel |
| Product Type | Inoculating Loop |
| Gauge | Standard |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido