Learn More
Bochem™ Chrome Steel PVC Coated 3-Fingers Clamps
Includes shaft. Bochem™ Chrome Steel PVC Coated 3-Fingers Clamps span width independently adjustable by 2 regulating screws.
Brand:  Bochem™ 5626
 View more versions of this product
Product Code. 10556951
  • 39.05 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Finger PVC coated
  • Span width independently adjustable by 2 regulating screws
  • With shaft
Specifications
Specifications
1 Ea.
Chrome Steel
0 to 35 mm
3 Finger Clamp
150 mm
10 mm